Learn More
Azzota Corp Honey refract 13-25pct ATC
Catalog No. 5000887307
Encompass_Preferred
RHH13-25T Heavy-Duty Portable Refractometer for Honey Test Sturdy design made of Aluminum Honey Moisture range 13-25% resolution 0 1% With ATC The line is pre-setup at Brix 19 6%
Catalog No. 50-008-87307 Supplier Azzota Corp Supplier No. RHH1325T
RHH13-25T Heavy-Duty Portable Refractometer for Honey Test Sturdy design made of Aluminum Honey Moisture range 13-25% resolution 0 1% With ATC The line is pre-setup at Brix 19 6%