Learn More
Azzota Corp Honey refract 58-92pct ATC
Catalog No. 5000887275
Encompass_Preferred
RHH58-92T Heavy-Duty Portable Refractometer for Honey Test Sturdy design made of Aluminum Brix Range 58-92% Baume 38-43Be Water 12-27% With ATC
Catalog No. 50-008-87275 Supplier Azzota Corp Supplier No. RHH5892T
RHH58-92T Heavy-Duty Portable Refractometer for Honey Test Sturdy design made of Aluminum Brix Range 58-92% Baume 38-43Be Water 12-27% With ATC