Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Barricade Tape

Catalog No. 19120633
Change view
Click to view available options
Color:
Red
Displayed Text:
Caution
Length (Metric):
91.4 m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Displayed Text Length (Metric)
19-120-633 Red Caution 91.4 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-120-633 Supplier Honeywell™ Supplier No. CT3RE9/300N

missing translation for 'validMainframeMsg'

Dispenses quickly and easily to effectively designate a hazardous area

  • Polyethylene tape with repeating message (Danger Do Not Enter, Caution Do Not Enter, Caution Buried Cable, Danger or Caution)
  • Width: 7.62cm (3 in.)
  • Comes in convenient dispenser box
TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W) 300 ft. x 3 in.
Material HDPE
Width (Metric) 7.62 cm
Color Red
Width (English) 3 in.
Thickness 0.002 mil
Length (English) 300 ft.
For Use With (Application) Dispenses quickly and easily to effectively designate a hazardous area
Length (Metric) 91.4 m
Type Barricade Tape
Displayed Text Caution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.