Learn More
Honeywell™ Ben 2 Plus Proximity Helmet
Catalog No. 19167106
Change view
Click to view available options
Material:
J-Fire military proximity shroud/ear cover
Military proximity shroud/ear cover
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-167-106 Military proximity shroud/ear cover
19-167-107 J-Fire military proximity shroud/ear cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-167-106 Supplier Honeywell™ Supplier No. SAEGSA00503
Add to Cart
Add to Cart
For proximity/ARFF operations and specialized military applications
  • 6 × 0.130 in. gold coat faceshield
  • Nomex™ Quick Release chinstrap with postman slide
  • Military proximity shroud/ear cover
  • Kevlar™ felt winter liner dome
  • Eagle front holder—golden alloy plating aluminum with hardware
Certifications
Meet NFPA 1971, 2007 Edition.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets NFPA 1971, 2007 Edition.
Color Yellow
Liner Material Dupont™ Kevlar™ aramid fiber Felt Winter
Material Military proximity shroud/ear cover