Learn More
Honeywell™ Bull Dog™ Leather Palm Cut Resistant Gloves
Description
- Aramid fiber shell is 5 times stronger than steel on an equal weight basis
- Enhanced grip capabilities in wet or oily applications
- Reinforced leather thumb crotch provides additional protection in this high-wear area
- Complete protection with split leather on palms and wrapped between fingers
Specifications
Specifications
| Color | Gray, Yellow |
| Hand | Specific |
| Material | Aramid Fiber, Leather |
| Product Type | Cut-resistant Gloves |
| Size | Universal |
| Glove Cuff Color | Yellow |
| Glove Cuff Style | Knit |
| Palm Type | Leather |
| Series | Bull Dog™ |