Learn More
MSC Honeywell BW IntelliDoX Enabler Kit, DX-ENBL-NA, 1 ea
Catalog No. 5000765550
Encompass
The BW IntelliDoX Enabler Kit is an add-on designed to assist the BW IntelliDoX Docking Station in its function. This kit is not capable of independent operation.
  • Includes power supply with region-specific AC power cord, ethernet cable, inlet (purge) filter assembly, 3 ft. (1 m) calibration gas tubing, 3 ft. (1 m) purge gas tubing, 15 ft. (4.5 m) exhaust tubing, quick connect fittings, and an IntelliDoX inlet end plate.
  • Compatible with IntelliDoX models for GasAlertMicroClip XL or X3 (DX-MCLP), BW Clip4 (DX-BWC4), BW Clip Series (DX-CLIP), BW Icon Series and Flex (DX-CP), and Gas Alert Max XT II (DX-MAXXT).
Catalog No. 50-007-65550 Supplier MSC Supplier No. 35925544
The BW IntelliDoX Enabler Kit is an add-on designed to assist the BW IntelliDoX Docking Station in its function. This kit is not capable of independent operation.
  • Includes power supply with region-specific AC power cord, ethernet cable, inlet (purge) filter assembly, 3 ft. (1 m) calibration gas tubing, 3 ft. (1 m) purge gas tubing, 15 ft. (4.5 m) exhaust tubing, quick connect fittings, and an IntelliDoX inlet end plate.
  • Compatible with IntelliDoX models for GasAlertMicroClip XL or X3 (DX-MCLP), BW Clip4 (DX-BWC4), BW Clip Series (DX-CLIP), BW Icon Series and Flex (DX-CP), and Gas Alert Max XT II (DX-MAXXT).