Learn More
MSC Honeywell BW MicroClip X3 Multi-Gas Detector, 4-Gas, IP68, 1 ea
Catalog No. 5000685863
Encompass
The Honeywell BW MicroClip X3 multi-gas portable detector offers reliable, cost-effective, and user-friendly monitoring for up to four gases. It features an IP68 rating for water protection and a guaranteed runtime of at least 12 hours in cold weather.
  • Simultaneously monitors for up to four gases (O2, H2S, CO, and combustibles)
  • Battery runtime of 18 hours in normal temperatures
  • Guaranteed 12-hour runtime in cold weather (-4°F / -20°C)
  • IP68 rating for unmatched water protection
  • Compact, lightweight, and easy to use with one-button operation
  • Flashing green IntelliFlash light for compliance at a glance
  • Reflex Technology ensures sensor responsiveness
  • Extended reliability with a 3-year warranty
  • Compatible with IntelliDox Docking Station for automated bump tests and calibration
  • Device management with SafetySuite
Catalog No. 50-006-85863 Supplier MSC Supplier No. 46487526
The Honeywell BW MicroClip X3 multi-gas portable detector offers reliable, cost-effective, and user-friendly monitoring for up to four gases. It features an IP68 rating for water protection and a guaranteed runtime of at least 12 hours in cold weather.
  • Simultaneously monitors for up to four gases (O2, H2S, CO, and combustibles)
  • Battery runtime of 18 hours in normal temperatures
  • Guaranteed 12-hour runtime in cold weather (-4°F / -20°C)
  • IP68 rating for unmatched water protection
  • Compact, lightweight, and easy to use with one-button operation
  • Flashing green IntelliFlash light for compliance at a glance
  • Reflex Technology ensures sensor responsiveness
  • Extended reliability with a 3-year warranty
  • Compatible with IntelliDox Docking Station for automated bump tests and calibration
  • Device management with SafetySuite