Learn More
MSC Honeywell BW Multi-Gas Detector, 4-Gas, Alkaline Version, 1 ea
Catalog No. 5000684354
Encompass
The Honeywell BW GasAlertQuattro multi-gas detector is a rugged, reliable device for personal gas detection. It features IntelliFlash™ for continuous visual compliance verification and detects up to four gases, suitable for industrial applications including confined space entry.
  • IntelliFlash verifies operation and compliance
  • Detects hydrogen sulfide, carbon monoxide, oxygen, and combustible gases
  • Compact, lightweight design with built-in concussion-proof boot
  • Compatible with MicroDock II automatic test and calibration system
  • Full function self-test on start-up
  • One-button operation and straightforward user interface
  • Comprehensive data and event logging
  • Field-proven SureCell sensors
  • Multi-language support
  • IP66/IP67 rated
Catalog No. 50-006-84354 Supplier MSC Supplier No. 50511039
The Honeywell BW GasAlertQuattro multi-gas detector is a rugged, reliable device for personal gas detection. It features IntelliFlash™ for continuous visual compliance verification and detects up to four gases, suitable for industrial applications including confined space entry.
  • IntelliFlash verifies operation and compliance
  • Detects hydrogen sulfide, carbon monoxide, oxygen, and combustible gases
  • Compact, lightweight design with built-in concussion-proof boot
  • Compatible with MicroDock II automatic test and calibration system
  • Full function self-test on start-up
  • One-button operation and straightforward user interface
  • Comprehensive data and event logging
  • Field-proven SureCell sensors
  • Multi-language support
  • IP66/IP67 rated