Learn More
MSC Honeywell BW Multi-Gas Detector, 4 Gas, Integrated Pump, 1 ea
Catalog No. 5000687502
Encompass
The Honeywell BW™ Max XT II is a smart, simple, and economical multi-gas detector for enhanced worker safety. It features one-button operation with a robust, motorized SmartSample pump for intelligent, remote sampling, suitable for confined spaces. This device reliably monitors up to four hazards and is compatible with the MicroDock II automated test and calibration system.
  • Monitors H2S, CO, O2, and combustibles.
  • Integrated sampling pump with reliable diaphragm technology.
  • Continuous LCD shows real-time gas concentrations.
  • Audible (95 dB), vibrating, and visual alarms.
  • Built-in concussion-proof boot.
  • IP66/67 ingress protection.
  • Standard datalogging and event logging.
Catalog No. 50-006-87502 Supplier MSC Supplier No. 50668631
The Honeywell BW™ Max XT II is a smart, simple, and economical multi-gas detector for enhanced worker safety. It features one-button operation with a robust, motorized SmartSample pump for intelligent, remote sampling, suitable for confined spaces. This device reliably monitors up to four hazards and is compatible with the MicroDock II automated test and calibration system.
  • Monitors H2S, CO, O2, and combustibles.
  • Integrated sampling pump with reliable diaphragm technology.
  • Continuous LCD shows real-time gas concentrations.
  • Audible (95 dB), vibrating, and visual alarms.
  • Built-in concussion-proof boot.
  • IP66/67 ingress protection.
  • Standard datalogging and event logging.