Learn More
MSC Honeywell BW Multi-gas Detector, 5-Year O2 Sensor, IP68, 1 ea
Catalog No. 5000676294
Encompass
The Honeywell BW MicroClip X3 multi-gas detector provides continuous monitoring for up to four gas hazards. This compact, wearable device ensures safety in hazardous environments with its reliable performance, long-life oxygen sensor, and user-friendly operation.
  • Five-year life expectancy for all sensors
  • Three-year warranty covering all sensors
  • New oxygen sensor with a five-year lifespan
  • IP68 rated for maximum protection from dust and water
  • Guaranteed 12-hour runtime at -20°C
  • One-button, tamper-proof operation
  • Real-time gas concentrations shown on LCD
  • IntelliFlash light verifies operation and compliance
  • Compatible with IntelliDoX docking station
Catalog No. 50-006-76294 Supplier MSC Supplier No. 42289173
The Honeywell BW MicroClip X3 multi-gas detector provides continuous monitoring for up to four gas hazards. This compact, wearable device ensures safety in hazardous environments with its reliable performance, long-life oxygen sensor, and user-friendly operation.
  • Five-year life expectancy for all sensors
  • Three-year warranty covering all sensors
  • New oxygen sensor with a five-year lifespan
  • IP68 rated for maximum protection from dust and water
  • Guaranteed 12-hour runtime at -20°C
  • One-button, tamper-proof operation
  • Real-time gas concentrations shown on LCD
  • IntelliFlash light verifies operation and compliance
  • Compatible with IntelliDoX docking station