Learn More
LESMAN INSTRUMENT COMPANY 10-5/16" 7D Circular Chart, 100/box
Catalog No. NC9802250
Encompass_Preferred
Honeywell Circular Chart | Chart Size (Diam): 10-5/16-Inch | 7 Day Rotation | Range: -5 to 50 | Compatible Recorders: AR100, DR4200, DR4300 | Box of 100.
Catalog No. NC9802250 Supplier LESMAN INSTRUMENT COMPANY Supplier No. 24001661022
Honeywell Circular Chart | Chart Size (Diam): 10-5/16-Inch | 7 Day Rotation | Range: -5 to 50 | Compatible Recorders: AR100, DR4200, DR4300 | Box of 100.