Learn More
MSC Honeywell Chemical Resistant Gloves, Butyl, 13 mil, Size 10, 11 in Length
Catalog No. 5000508558
Encompass
These Butyl chemical resistant gloves provide superior protection against a wide range of aggressive chemicals, including ketones and esters, solvents, acids, and bases. They offer high permeation resistance to gas and water vapor, ensuring safety while maintaining flexibility and dexterity for the user.
  • Unsupported Butyl construction with a smooth finish.
  • Rolled edge cuff reinforces tear resistance and prevents drips.
  • Remains supple from -30°C up to +50°C.
  • Curved finger and hand design ensures a comfortable, ergonomic fit.
  • Excellent resistance to many solvents, acids, medium concentrated bases, ozone, and heat.
  • Highest permeation resistance to gas or water vapors.
  • Sanitized interior lining for enhanced hygiene.
  • Good resistance to tear.
Catalog No. 50-005-08558 Supplier MSC Supplier No. 01904069
These Butyl chemical resistant gloves provide superior protection against a wide range of aggressive chemicals, including ketones and esters, solvents, acids, and bases. They offer high permeation resistance to gas and water vapor, ensuring safety while maintaining flexibility and dexterity for the user.
  • Unsupported Butyl construction with a smooth finish.
  • Rolled edge cuff reinforces tear resistance and prevents drips.
  • Remains supple from -30°C up to +50°C.
  • Curved finger and hand design ensures a comfortable, ergonomic fit.
  • Excellent resistance to many solvents, acids, medium concentrated bases, ozone, and heat.
  • Highest permeation resistance to gas or water vapors.
  • Sanitized interior lining for enhanced hygiene.
  • Good resistance to tear.