Learn More
MSC Honeywell Chemical Resistant Gloves, Nitrile, 40 Mil, 12 in
Catalog No. 5000507905
Encompass
These chemical resistant gloves are designed for high-performance applications, offering superior resistance to cuts, snags, abrasions, and punctures. Constructed with nitrile over an interlock knitted liner, these 40 mil thick gloves provide high resistance to solvents, caustics, and animal fats, making them suitable for food processing and chemical handling.
  • Offers superior resistance to cuts, snags, abrasions, and punctures.
  • Provides high resistance to solvents, caustics, and animal fats.
  • Features a soft and comfortable interlock knit liner.
  • Treated with an anti-bacterial and anti-mildew agent.
  • Curved shape for natural fit and reduced worker hand fatigue.
  • Meets USDA and FDA regulations for food processing.
  • Available in sizes 7-11.
  • Unlined for comfort.
  • Rough texture for enhanced grip.
  • Latex free.
  • 12 inch length.
Catalog No. 50-005-07905 Supplier MSC Supplier No. 56099898
These chemical resistant gloves are designed for high-performance applications, offering superior resistance to cuts, snags, abrasions, and punctures. Constructed with nitrile over an interlock knitted liner, these 40 mil thick gloves provide high resistance to solvents, caustics, and animal fats, making them suitable for food processing and chemical handling.
  • Offers superior resistance to cuts, snags, abrasions, and punctures.
  • Provides high resistance to solvents, caustics, and animal fats.
  • Features a soft and comfortable interlock knit liner.
  • Treated with an anti-bacterial and anti-mildew agent.
  • Curved shape for natural fit and reduced worker hand fatigue.
  • Meets USDA and FDA regulations for food processing.
  • Available in sizes 7-11.
  • Unlined for comfort.
  • Rough texture for enhanced grip.
  • Latex free.
  • 12 inch length.