Learn More
MSC Honeywell Chemical Resistant PVC Glove, 12 in, Triple-Dipped, Pinked Edge Cuff
Catalog No. 5000508258
Encompass
This Honeywell PowerCoat 660 triple-dipped PVC glove protects hands from hazardous liquids and chemicals. It features a soft, interlock knit liner and a pinked edge cuff, providing comfort and flexibility while resisting oils, chemicals, acids, and solvents. It also serves as an excellent latex alternative for allergy sufferers.
  • Protects hands from hazardous liquids and chemicals.
  • Triple-dipped PVC construction.
  • 12-inch length.
  • Soft, interlock knit liner provides comfort and ventilation.
  • Pinked edge cuff.
  • Resists oils, chemicals, acids, and solvents.
  • Flexible design reduces hand fatigue.
  • Seamless liner prevents rubbing.
  • Combines latex-like suppleness with PVC chemical resistance.
  • Ideal for individuals with latex allergies.
  • Conforms to EN388 for abrasion and cut resistance.
Catalog No. 50-005-08258 Supplier MSC Supplier No. 62097035
This Honeywell PowerCoat 660 triple-dipped PVC glove protects hands from hazardous liquids and chemicals. It features a soft, interlock knit liner and a pinked edge cuff, providing comfort and flexibility while resisting oils, chemicals, acids, and solvents. It also serves as an excellent latex alternative for allergy sufferers.
  • Protects hands from hazardous liquids and chemicals.
  • Triple-dipped PVC construction.
  • 12-inch length.
  • Soft, interlock knit liner provides comfort and ventilation.
  • Pinked edge cuff.
  • Resists oils, chemicals, acids, and solvents.
  • Flexible design reduces hand fatigue.
  • Seamless liner prevents rubbing.
  • Combines latex-like suppleness with PVC chemical resistance.
  • Ideal for individuals with latex allergies.
  • Conforms to EN388 for abrasion and cut resistance.