Learn More
Graphic Controls LLC HONEYWELL CIRCULAR CHARTS100PK
Catalog No. NC3039330
Encompass_Preferred
Chart Size10.313"Time Rotation24HRanges0 - 300 Thermocouple 0 TO 300 ºC J T/C (50 TO 250 BY 50
Catalog No. NC3039330 Supplier Graphic Controls LLC Supplier No. 20538817
Chart Size10.313"Time Rotation24HRanges0 - 300 Thermocouple 0 TO 300 ºC J T/C (50 TO 250 BY 50