Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Survivair Panther™ / Cougar™ SCBA Accessory, Skirt Assembly

Catalog No. 17377613
Change view
Click to view available options
Size: Standard; Color: Black; Includes headstrap, metal rim, nut, screw, lens and skirt with buckles
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-377-613 Size: Standard; Color: Black; Includes headstrap, metal rim, nut, screw, lens and skirt with buckles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-377-613 Supplier Honeywell™ Supplier No. 968422

Specifications

Color Black
Applications Firefighting
For Use With (Equipment) Survivair Panther, Cougar SCBA
Model 968422
Product Type Skirt Assembly
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.