Learn More
MSC Honeywell Cut, Puncture & Abrasion-Resistant Gloves, Size 11, Cut A2, 18 Gauge
Catalog No. 5000510024
Encompass
These cut, puncture, and abrasion-resistant gloves feature a lightweight 18 gauge liner made from HPPE, Nylon, Carbon, and Elastane. They are coated with polyurethane on the palm and fingertips for enhanced grip and protection. Designed for comfort and dexterity, these gloves are also antistatic, touchscreen capable, and food certified.
  • Antistatic properties certified to EN 16350
  • Improved liners with a high 18 gauge count
  • Touch screen capability for device use
  • Silicone-free for automotive applications
  • Color-coded marking for cut protection levels
  • Low DMF content (less than 100 ppm)
  • OEKO-TEX 100 certified for food handling
  • Washable (3 cycles)
  • Glass fibre-free material
  • Optimum grip performance
  • Seamless lining for comfort
Catalog No. 50-005-10024 Supplier MSC Supplier No. 05764220
These cut, puncture, and abrasion-resistant gloves feature a lightweight 18 gauge liner made from HPPE, Nylon, Carbon, and Elastane. They are coated with polyurethane on the palm and fingertips for enhanced grip and protection. Designed for comfort and dexterity, these gloves are also antistatic, touchscreen capable, and food certified.
  • Antistatic properties certified to EN 16350
  • Improved liners with a high 18 gauge count
  • Touch screen capability for device use
  • Silicone-free for automotive applications
  • Color-coded marking for cut protection levels
  • Low DMF content (less than 100 ppm)
  • OEKO-TEX 100 certified for food handling
  • Washable (3 cycles)
  • Glass fibre-free material
  • Optimum grip performance
  • Seamless lining for comfort