These cut-resistant gloves offer a balanced solution for workers needing both protection and dexterity. Constructed with a 15-gauge nylon liner and a nitrile micro-foam coating, they provide enhanced cut resistance, comfort, and a secure grip for extended wear. The gloves are designed for a wide range of applications, ensuring simplified hand protection.
- Provides excellent grip in wet, dry, and slightly oily conditions.
- Touchscreen compatible.
- Reinforced thumb crotch for enhanced durability.
- Washable for reduced cost of ownership.
- Lightweight 15 gauge nylon liner offers high breathability and tactile sensitivity.
- Micro-foam nitrile coating provides 360° breathability and enhanced durability.
- Designed to prevent hand fatigue and reduce allergies.
- Ensures essential protection against minor abrasions and sharp edges.
- Longer cuff provides wrist protection.