Learn More
MSC Honeywell Cut Resistant Glove, HPPE/Basalt, Nitrile Palm, ANSI Cut A4, Size Medium
Catalog No. 5000509584
Encompass
The Honeywell CoreShield™ Cut Resistant Gloves are engineered to provide optimal hand protection in various industrial applications. Featuring a 13-gauge HPPE/basalt liner and a smooth nitrile coating on the palm and fingertips, these gloves offer enhanced cut, puncture, and abrasion resistance. Designed for user comfort and ease of selection, they balance protection with cost-effectiveness.
  • Enhanced cut resistance
  • Comfortable and easy to select
  • Cost-effective hand protection
  • Touchscreen compatible
  • Reinforced thumb crotch
  • Silicone free
  • Washable design
  • Dry grip performance
  • Liquid repellent and water/oil proof coating
  • Seamless lining
Catalog No. 50-005-09584 Supplier MSC Supplier No. 94865458
The Honeywell CoreShield™ Cut Resistant Gloves are engineered to provide optimal hand protection in various industrial applications. Featuring a 13-gauge HPPE/basalt liner and a smooth nitrile coating on the palm and fingertips, these gloves offer enhanced cut, puncture, and abrasion resistance. Designed for user comfort and ease of selection, they balance protection with cost-effectiveness.
  • Enhanced cut resistance
  • Comfortable and easy to select
  • Cost-effective hand protection
  • Touchscreen compatible
  • Reinforced thumb crotch
  • Silicone free
  • Washable design
  • Dry grip performance
  • Liquid repellent and water/oil proof coating
  • Seamless lining