Learn More
MSC Honeywell Cut-Resistant Gloves, Cut A6, 13 Gauge, Medium
Catalog No. 5000507716
Encompass
These gloves offer superior cut, puncture, and abrasion resistance, designed for comfort and extended wear in industrial settings. Featuring a 13-gauge HPPE liner and a micro-foam nitrile coating, they provide excellent dexterity and grip while meeting rigorous safety standards.
  • Lightweight design prevents hand fatigue.
  • Skin-friendly yarns reduce allergy and improve dexterity.
  • Micro-foam nitrile coating provides 360° breathability, grip, and abrasion resistance.
  • Reinforced thumb for extended product life.
  • Washable, reducing cost of ownership.
  • Longer cuff offers wrist protection.
  • Touchscreen compatible.
  • 13-gauge construction for great fit and dexterity.
  • Color-coded system for easy selection.
  • Certified by OEKO-TEX.
Catalog No. 50-005-07716 Supplier MSC Supplier No. 90740325
These gloves offer superior cut, puncture, and abrasion resistance, designed for comfort and extended wear in industrial settings. Featuring a 13-gauge HPPE liner and a micro-foam nitrile coating, they provide excellent dexterity and grip while meeting rigorous safety standards.
  • Lightweight design prevents hand fatigue.
  • Skin-friendly yarns reduce allergy and improve dexterity.
  • Micro-foam nitrile coating provides 360° breathability, grip, and abrasion resistance.
  • Reinforced thumb for extended product life.
  • Washable, reducing cost of ownership.
  • Longer cuff offers wrist protection.
  • Touchscreen compatible.
  • 13-gauge construction for great fit and dexterity.
  • Color-coded system for easy selection.
  • Certified by OEKO-TEX.