Learn More
MSC Honeywell Cut-Resistant Gloves, Dyneema, PU Coated, Cut Level A2, Knit Wrist
Catalog No. 5000509120
Encompass
These cut-resistant gloves offer reliable hand protection for various industrial applications. Constructed with Dyneema material and featuring a polyurethane coating, they are designed for durability and a secure grip. These gloves meet ANSI/ISEA Cut Level A2 and provide abrasion resistance, making them suitable for tasks requiring protection against cuts and wear.
  • Cut-resistant with ANSI/ISEA A2 level for enhanced safety.
  • Dyneema material provides high strength and cut protection.
  • Polyurethane (PU) coating offers a rough finish for improved grip.
  • 13 gauge thickness for dexterity and protection.
  • Knit wrist cuff style ensures a comfortable and secure fit.
  • Silicone-free construction.
  • Certified to ANSI/ISEA 105:2016 and EN 388:2016 standards.
  • Abrasion level 6 and puncture level 2 for comprehensive protection.
Catalog No. 50-005-09120 Supplier MSC Supplier No. 97365696
These cut-resistant gloves offer reliable hand protection for various industrial applications. Constructed with Dyneema material and featuring a polyurethane coating, they are designed for durability and a secure grip. These gloves meet ANSI/ISEA Cut Level A2 and provide abrasion resistance, making them suitable for tasks requiring protection against cuts and wear.
  • Cut-resistant with ANSI/ISEA A2 level for enhanced safety.
  • Dyneema material provides high strength and cut protection.
  • Polyurethane (PU) coating offers a rough finish for improved grip.
  • 13 gauge thickness for dexterity and protection.
  • Knit wrist cuff style ensures a comfortable and secure fit.
  • Silicone-free construction.
  • Certified to ANSI/ISEA 105:2016 and EN 388:2016 standards.
  • Abrasion level 6 and puncture level 2 for comprehensive protection.