This multi-gas portable detector is part of the MicroClip Series, known for its long runtime, reliability, small size, and ease of use. The X3 model features a new O2 sensor and an extended warranty, offering enhanced reliability and user-friendliness.
- Monitors up to four gases simultaneously: O2, H2S, CO, and combustibles
- Battery runtime of 18 hours in normal temperatures, with at least 12 hours guaranteed in cold weather
- Enhanced water protection with IP68 rating
- Extended 3-year warranty for reliability
- Guaranteed to operate an entire shift, even at -4°F (-20°C)
- Compact, lightweight, and user-friendly with one-button operation
- Compatible with Safety Suite for device management
- IntelliFlash™ light provides compliance at a glance
- Reflex™ Technology ensures sensor responsiveness