Learn More
MSC Honeywell Disposable Gloves, Nitrile, 5 mil, Blue, Small, Powder-Free
Catalog No. 5000508343
Encompass
These disposable nitrile gloves offer protection against chemical hazards and abrasion. They provide excellent puncture and tear resistance, making them suitable for various industrial and service applications. The gloves comply with USDA and FDA regulations, 21 CFR, for use in food processing.
  • Nitrile formulation helps prevent latex allergic reactions.
  • Provides excellent puncture and tear resistance.
  • Offers better chemical resistance than latex and vinyl.
  • Rolled cuff helps prevent tearing during donning and doffing.
  • Smooth surface finish.
  • Ambidextrous design for easy wear.
  • Powder-free for sensitive environments.
  • Exam grade for quality assurance.
  • Unlined for enhanced dexterity.
Catalog No. 50-005-08343 Supplier MSC Supplier No. 54641147
These disposable nitrile gloves offer protection against chemical hazards and abrasion. They provide excellent puncture and tear resistance, making them suitable for various industrial and service applications. The gloves comply with USDA and FDA regulations, 21 CFR, for use in food processing.
  • Nitrile formulation helps prevent latex allergic reactions.
  • Provides excellent puncture and tear resistance.
  • Offers better chemical resistance than latex and vinyl.
  • Rolled cuff helps prevent tearing during donning and doffing.
  • Smooth surface finish.
  • Ambidextrous design for easy wear.
  • Powder-free for sensitive environments.
  • Exam grade for quality assurance.
  • Unlined for enhanced dexterity.