Learn More
MSC Honeywell Earmuff, Helmet Capmount, NRR 21
Catalog No. 5000509422
Encompass
Hearing protection devices are only effective when they are worn, so comfort is essential. These passive earmuffs are designed to provide optimum comfort over long periods of use. They fit a wide range of head sizes, have a wide cup opening with memory foam ear cushions, and a lightweight cushioned headband, making them solutions users will want to wear.
  • Hard hat cap mounted
  • Noise Reduction Rating (NRR) of 21
  • Lightweight cushioned headband with steel-wire construction
  • Memory foam ear cushions for greater comfort and attenuation
  • Robust construction with oil-resistant cup surface
  • Air Flow Control technology for optimum attenuation
  • Adjustable headband
  • Non-conductive design
  • Wide cup openings for larger ears or hearing aids
  • Designed for compatibility with other Personal Protective Equipment (PPE)
Catalog No. 50-005-09422 Supplier MSC Supplier No. 95460630
Hearing protection devices are only effective when they are worn, so comfort is essential. These passive earmuffs are designed to provide optimum comfort over long periods of use. They fit a wide range of head sizes, have a wide cup opening with memory foam ear cushions, and a lightweight cushioned headband, making them solutions users will want to wear.
  • Hard hat cap mounted
  • Noise Reduction Rating (NRR) of 21
  • Lightweight cushioned headband with steel-wire construction
  • Memory foam ear cushions for greater comfort and attenuation
  • Robust construction with oil-resistant cup surface
  • Air Flow Control technology for optimum attenuation
  • Adjustable headband
  • Non-conductive design
  • Wide cup openings for larger ears or hearing aids
  • Designed for compatibility with other Personal Protective Equipment (PPE)