Learn More
Honeywell™ Eclipse™ 6400 Structural Firefighting Gloves
Catalog No. 19821895
Change view
Click to view available options
Size:
Large
2X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-821-895 Large
19-805-458 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-821-895 Supplier Honeywell™ Supplier No. GL6400LARG3
Add to Cart
Add to Cart
Innovative cuff design is convertible and can be worn as a knit wrist, or in a gauntlet style. DuPont™ Nomex™ fibers provide strength and durability, in addition to a high level of thermal protection.
  • Innovative cuff is convertible and can be worn as either a knit wrist or gauntlet cuff
  • Extra-wide cuff, constructed of 2-ply Nomex™ spandex makes donning easy
  • DuPont™ Nomex™ fibers provide superior strength and durability
  • Anti-slash strips provide an extra band of protection to the palm and first knuckle
  • Gloves provide a high level of thermal protection and are capable of standing up to high heat
  • Unique Swing Thumb™ bends in opposition to the fingertips, mimicking the motions of human hands
  • Crosstech™ Moisture Barrier keeps hands dry and comfortable
  • 360° inner pull tab for additional strength in tough applications
Certifications
NFPA 1971
TRUSTED_SUSTAINABILITY
Specifications
Back Material Elkhide Leather
Certifications/Compliance NFPA 1971
Cuff Material Nomex™ Spandex
Coating Material Uncoated
Color Brown, Dark Blue
Hand Specific
Product Type Structural Firefighting Gloves
Size Large
Glove Cuff Color White
Glove Cuff Style Gauntlet
Includes Crosstech™ Moisture Barrier
Liner Material Self Extinguishing Fleece (SEF)
Palm Type Cowhide Leather
Series GL-6400 Eclipse