Learn More
Honeywell™ Eclipse™ 8700 Structural Firefighting Gloves
Catalog No. 1980540008
Change view
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-805-40008 3X-Large
19-805-40001 2X-Small
19-805-40002 X-Small
19-805-40003 Small
19-805-40004 Medium
19-805-40005 Large
19-805-40006 X-Large
19-805-40007 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-805-40008 Supplier Honeywell™ Supplier No. GL87003XL
Add to Cart
Add to Cart
Innovative cuff design is convertible and can be worn as a knit wrist, or in a gauntlet style. DuPont™ Nomex™ fibers provide strength and durability, in addition to a high level of thermal protection.
  • Innovative cuff is convertible and can be worn as either a knit wrist or gauntlet cuff
  • Extra-wide cuff, constructed of 2-ply Nomex™ spandex makes donning easy
  • DuPont™ Nomex™ fibers provide superior strength and durability
  • Anti-slash strips provide an extra band of protection to the palm and first knuckle
  • Gloves provide a high level of thermal protection and are capable of standing up to high heat
  • Unique Swing Thumb™ bends in opposition to the fingertips, mimicking the motions of human hands
  • Pyrotech™ Moisture Barrier keeps hands dry and comfortable
  • 360° inner pull tab for additional strength in tough applications
Certifications
NFPA 1971-2007
TRUSTED_SUSTAINABILITY
Specifications
Back Material Elkhide Leather
Certifications/Compliance NFPA 1971-2007
Cuff Material Nomex™ Spandex
Coating Material Uncoated
Color Dark Blue, Yellow
Hand Specific
Product Type Structural Firefighting Gloves
Size 3X-Large
Glove Cuff Color White
Glove Cuff Style Gauntlet
Includes Pyrotect™ Moisture Barrier
Liner Material Nomex™/Aramid Fiber
Palm Type Kangaroo Leather
Series GL-8700 Eclipse