Learn More
Enterprise Technology Solutions Honeywell Faceshield Replacement Visor, 10/BG, CL
SDP
Supplier:  Enterprise Technology Solutions HWLA815040
Encompass_Preferred
Faceshield replacement visor features an anti-scratch coating that protects the surface from damage for long-lasting clarity. 99.9 percent ultraviolet protection helps protect from eye damage for safe outdoor use. Curved construction provides extra space to allow you to wear safety glasses or respirators. Replacement visor is designed for Honeywell faceshields (sold separately).
Catalog No. 50-008-28244