Learn More
MSC Honeywell Faceshield Visor, Propionate, Clear, 8.5 x 15 x .07 in, ANSI Z87+
Catalog No. 5000510023
Encompass
This Honeywell Protecto-Shield replacement faceshield visor is constructed from tough propionate and designed for high-impact applications. It features a universal hole pattern for broad compatibility with various headgear and provides splash resistance, ensuring worker safety in diverse environments.
  • Heat resistant for various applications.
  • Easy attachment with no snaps, posts, or levers.
  • Universal hole pattern for compatibility with various headgear.
  • Splash resistant design with a chemical drip edge.
  • Reinforced edge resists cracking.
  • Meets ANSI Z87+ (high impact) standard.
  • Tested to meet CSA Z94.3 standard.
Catalog No. 50-005-10023 Supplier MSC Supplier No. 05158266
This Honeywell Protecto-Shield replacement faceshield visor is constructed from tough propionate and designed for high-impact applications. It features a universal hole pattern for broad compatibility with various headgear and provides splash resistance, ensuring worker safety in diverse environments.
  • Heat resistant for various applications.
  • Easy attachment with no snaps, posts, or levers.
  • Universal hole pattern for compatibility with various headgear.
  • Splash resistant design with a chemical drip edge.
  • Reinforced edge resists cracking.
  • Meets ANSI Z87+ (high impact) standard.
  • Tested to meet CSA Z94.3 standard.