Learn More
MSC Honeywell Fendall Eye Wash Station, Portable, Sterile, 0.4 GPM, 15 Min Duration
Catalog No. 5000508715
Encompass
This eye wash station is a portable, sterile system that provides 15 minutes of uninterrupted flushing with 100% sterile saline solution. It delivers 0.4 GPM, meeting ANSI Z358.1-2014 standards for primary eyewash stations. The unit operates with quick-change cartridges and features an alarm system for proper use.
  • Uses quick-change 6.87 gallon (26 L) cartridges
  • Alarm sounds at the start and end of activation
  • Features an arm/headrest for user comfort
  • Requires no plumbing for installation
  • Sleek assembly allows for easy placement near hazards
  • Includes easy-view side windows to check solution expiration date
Catalog No. 50-005-08715 Supplier MSC Supplier No. 65372856
This eye wash station is a portable, sterile system that provides 15 minutes of uninterrupted flushing with 100% sterile saline solution. It delivers 0.4 GPM, meeting ANSI Z358.1-2014 standards for primary eyewash stations. The unit operates with quick-change cartridges and features an alarm system for proper use.
  • Uses quick-change 6.87 gallon (26 L) cartridges
  • Alarm sounds at the start and end of activation
  • Features an arm/headrest for user comfort
  • Requires no plumbing for installation
  • Sleek assembly allows for easy placement near hazards
  • Includes easy-view side windows to check solution expiration date