Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Fendall Porta-Stream II Eyewash Station
SCI_ED

Catalog No. S47614A
Change view
Click to view available options
Capacity (English):
2048 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
S47614A 2048 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S47614A Supplier Honeywell™ Supplier No. 320002000000/EMD

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Convenient eyewash with no plumbing required.

The unequaled, original gravity-fed self-contained station

Check these features! No plumbing required. Unique hanging bracket permits easy installation on any flat horizontal or vertical surface. Exclusive full flow nozzles cover a wide facial area. Gravity fed—no moving parts. Simply remove the easy-to-locate nozzle strap to activate flushing—leaving the hands free to hold eyelids open. Locking fill-hole cap reduces risk of tampering with solution. Large fill hole located at top of unit simplifies refilling.

Order Info

Includes 180 oz. of Eyesaline™ concentrate which, when mixed with tap water, provides 16 gallons of isotonic solution as recommended by ANSI Z358.1-2009.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 2048 oz.
Capacity (Metric) 60566.58 mL
Certifications/Compliance ANSI Z358.1-2004
Includes Vinyl Eyewash Sign, Saline Concentrate
Material High Impact Polyethylene
Product Type Eyewash Station
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.