Learn More
MSC Honeywell Fibre-Metal Hard Hat, Thermoplastic, 8-Point Ratchet, Green
Catalog No. 5000509487
Encompass
This hard hat features a front brim style and an 8-point ratchet suspension for a secure fit. Constructed from thermoplastic, it provides protection as a Type 1, Class E hard hat, meeting ANSI Z89.1-2014 and CSA Z94.1-2015 standards. It accommodates head sizes from 6-1/2 to 8 and includes side slots for accessories.
  • Shell material: Thermoplastic
  • Suspension type: 8-point ratchet
  • Meets ANSI Type 1 and Class E standards
  • Fits head sizes 6-1/2 to 8
  • Includes side slots for accessories
  • Non-vented design
  • Green color
Catalog No. 50-005-09487 Supplier MSC Supplier No. 64302672
This hard hat features a front brim style and an 8-point ratchet suspension for a secure fit. Constructed from thermoplastic, it provides protection as a Type 1, Class E hard hat, meeting ANSI Z89.1-2014 and CSA Z94.1-2015 standards. It accommodates head sizes from 6-1/2 to 8 and includes side slots for accessories.
  • Shell material: Thermoplastic
  • Suspension type: 8-point ratchet
  • Meets ANSI Type 1 and Class E standards
  • Fits head sizes 6-1/2 to 8
  • Includes side slots for accessories
  • Non-vented design
  • Green color