Learn More
MSC Honeywell Fibre-Metal Headgear, Standard, Ratchet Adjustment
Catalog No. 5000509715
Encompass
The Fibre-Metal 3C Headgear is a standard, ratchet-adjustable headgear designed for comfort and durability. It features a deeply offset, pliable headband and free-floating arms, ensuring a secure and stable fit in various conditions.
  • Deeply offset, pliable headband designed to comfortably fit head contours.
  • Overhead band with multiple adjustments helps position the glassholder for the best view of the weld.
  • Exclusive free-floating arms balance and stabilize the helmet while minimizing pressure points.
  • Strip-proof ratchet head size adjustment provides small increment sizing for a secure fit.
  • Performs well in hot or cold conditions, resisting cracking, breaking, and aging.
Catalog No. 50-005-09715 Supplier MSC Supplier No. 56148224
The Fibre-Metal 3C Headgear is a standard, ratchet-adjustable headgear designed for comfort and durability. It features a deeply offset, pliable headband and free-floating arms, ensuring a secure and stable fit in various conditions.
  • Deeply offset, pliable headband designed to comfortably fit head contours.
  • Overhead band with multiple adjustments helps position the glassholder for the best view of the weld.
  • Exclusive free-floating arms balance and stabilize the helmet while minimizing pressure points.
  • Strip-proof ratchet head size adjustment provides small increment sizing for a secure fit.
  • Performs well in hot or cold conditions, resisting cracking, breaking, and aging.