Learn More
Honeywell™ 9550 Fire Mate™ Kangaroo Leather Firefighter Gloves
Catalog No. 19168245
Click to view available options
Size:
2X-Small
Small
Large
X-Large
3X-Large
Catalog No. 19-168-245
Supplier: Honeywell™ GL9550XXS
Only null left
Add to Cart
Add to Cart
Kangaroo leather on the palm absorbs significantly less water than traditional leather, while also providing a high level of abrasion resistance.
  • Kangaroo leather on the palm absorbs significantly less water than traditional leather
  • Pliable elkhide leather on the back of the glove is soft and comfortable
  • Crosstech™ Moisture Barrier is stable and durable with high temperature resistance
  • Nomex™/Aramid Fiber liner for thermal protection
  • Enhanced grip capabilities in wet applications
Certifications
NFPA 1971-2007, NFPA 1951-2007
TRUSTED_SUSTAINABILITY
Specifications
Back Material Elkhide Leather
Certifications/Compliance NFPA 1971-2007, NFPA 1951-2007
Cuff Material Leather
Coating Material Uncoated
Hand Specific
Product Type Structural Firefighting Gloves
Size 2X-Small
Glove Cuff Style Gauntlet
Includes Crosstech™ Moisture Barrier
Liner Material Nomex™/Aramid Fiber
Palm Type Kangaroo Leather
Series Fire Mate™ 9550
Thumb Style Wing
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.