Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ NorthFlex Light Task Plus II Cut-Resistant Gloves, Polyurethane Coated

Catalog No. 17805000
Change view
Click to view available options
Size:
Small
Medium
X-Large
2X-Large
Glove Cuff Color:
Blue
Orange
Red
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
17-805-000 Small Red
17-805-001 Medium Blue
17-805-003 X-Large White
17-805-004 2X-Large Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-805-000 Supplier Honeywell™ Supplier No. NFD15B/7S
This item is not returnable. View return policy

Lightweight Dyneema™ cut-resistant fiber gloves with white polyurethane palm and fingertip coating provides cut and abrasion protection.

  • Seamless, cut-resistant liner is made from Dyneema
  • Polyurethane palm coating is light for comfort and maximum tactility
  • High abrasion and puncture resistance
  • Glove conforms to the shape of the hand
  • Back of the glove is uncoated for ventilation and comfort
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Polyurethane
Color Black, Gray
Cut Resistance Rating ANSI A2
Hand Ambidextrous
Material Dyneema™
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Handling sharp objects, particularly sheet Metal and abrasives handling
Glove Cuff Color Red
Glove Cuff Style Knit
Hem Color Red
Model NFD15B/7S
Series NorthFlex Light Task Plus II™
Length (English) 4 in.
Length (Metric) 10.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.