Learn More
ORS Nasco HONEYWELL GLV CLNRM NIT 100/CS
Supplier:  ORS Nasco 068SK142W/9/HEM
Encompass_Preferred
Honeywell SK142W Cleanroom Gloves Nitrile, Size 9, Chemical Resistant, 10 per bag/10 bags per case. Powder Free, Bisque Finish
Catalog No. NC3073503