Learn More
Instrumart HONEYWELL INK WRTNG CIRC CHART
SDP
Catalog No. NC1342836
Encompass_Preferred
Honeywell 30755820 Ink Writing Circular Chart Range: 0 to 100 (Uniform Scale). Rotation: 24H / 7D. 25 per box Features Chart size: 10-5/16 Time rotation: 24H / 7D Number of ranges: 1 Range 1: 0 to 100 (Uniform Scale) 25 charts per box
Catalog No. NC1342836 Supplier Instrumart Supplier No. 30755820
Honeywell 30755820 Ink Writing Circular Chart Range: 0 to 100 (Uniform Scale). Rotation: 24H / 7D. 25 per box Features Chart size: 10-5/16 Time rotation: 24H / 7D Number of ranges: 1 Range 1: 0 to 100 (Uniform Scale) 25 charts per box