Learn More
MSC Honeywell Lens Cleaning Solution, 16 oz, Spray Bottle, Silicone-Free, Alcohol-Free
Catalog No. 5000509135
Encompass
This lens cleaning solution is specifically formulated for anti-fog and scratch resistant hardcoats. It does not contain silicone and is alcohol-free. It is available in a 16 oz refillable spray bottle.
  • Formulated for anti-fog and scratch-resistant hardcoats
  • Does not contain silicone
  • Alcohol-free
  • Refillable
  • Comes in a 16 oz spray bottle
Catalog No. 50-005-09135 Supplier MSC Supplier No. 96533252
This lens cleaning solution is specifically formulated for anti-fog and scratch resistant hardcoats. It does not contain silicone and is alcohol-free. It is available in a 16 oz refillable spray bottle.
  • Formulated for anti-fog and scratch-resistant hardcoats
  • Does not contain silicone
  • Alcohol-free
  • Refillable
  • Comes in a 16 oz spray bottle