Learn More
Honeywell™ Lite Force Plus Proximity Helmet
Catalog No. 19167105
Change view
Click to view available options
Material:
J-Fire military proximity shroud/ear cover
Military proximity shroud/ear cover
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-167-105 J-Fire military proximity shroud/ear cover
19-167-104 Military proximity shroud/ear cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-167-105 Supplier Honeywell™ Supplier No. SAEGSA00502
Add to Cart
Add to Cart
Strong, high-heat resistant shell and excellent fit
  • 6 × 0.130 in. gold coat faceshield
  • Nomex™ Quick Release chinstrap with postman slide
  • Proximity shroud/earcover
  • Five Reflexite™ lime bars, 1 × 4 in.
  • Kevlar™ felt winter liner dome
Certifications
NFPA 1971, 2007 Edition.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NFPA 1971, 2007 Edition.
Color Yellow
Liner Material Dupont™ Kevlar™ aramid fiber Felt Winter Liner
Material J-Fire military proximity shroud/ear cover