Learn More
MSC Honeywell MicroClip X3, Multi-Gas, IP68, 18 hr Battery, 1 ea
Catalog No. 5000677327
Encompass
The Honeywell BW MicroClip X3 gas detector is a compact, multi-gas portable detector known for its reliability, long battery life, and ease of use. It simultaneously monitors for up to four gases, including O2, H2S, CO, and combustibles (LEL), and features a three-year warranty and a five-year life expectancy.
  • Simultaneously monitors for up to four gases (O2, H2S, CO, and LEL)
  • Offers 18-hour battery runtime, with 12-hour cold weather guarantee
  • IP68 rated for maximum protection from dust and water
  • Compact, lightweight, and user-friendly one-button operation
  • Equipped with a durable, rechargeable lithium polymer battery
  • Features flashing green IntelliFlash light for compliance at a glance
  • Provides a 3-year warranty and 5-year life expectancy
Catalog No. 50-006-77327 Supplier MSC Supplier No. 42289140
The Honeywell BW MicroClip X3 gas detector is a compact, multi-gas portable detector known for its reliability, long battery life, and ease of use. It simultaneously monitors for up to four gases, including O2, H2S, CO, and combustibles (LEL), and features a three-year warranty and a five-year life expectancy.
  • Simultaneously monitors for up to four gases (O2, H2S, CO, and LEL)
  • Offers 18-hour battery runtime, with 12-hour cold weather guarantee
  • IP68 rated for maximum protection from dust and water
  • Compact, lightweight, and user-friendly one-button operation
  • Equipped with a durable, rechargeable lithium polymer battery
  • Features flashing green IntelliFlash light for compliance at a glance
  • Provides a 3-year warranty and 5-year life expectancy