Learn More
MSC Honeywell Miller Body Belt, Single D-Ring, X-Large, 1-3/4 in Webbing
Catalog No. 5000507964
Encompass
This body belt is designed for work positioning, featuring a single D-ring, X-Large size, and 1-3/4 inch nylon webbing with a tongue buckle. It is suitable for waist sizes from 42 to 52 inches and supports a maximum working load of 400 lb, meeting OSHA, ANSI, and CSA standards.
  • Designed for work positioning only.
  • Features a single D-ring.
  • Constructed with durable nylon webbing.
  • Equipped with a tongue buckle.
  • X-Large size, fitting waist sizes from 42 to 52 inches.
  • Webbing width of 1-3/4 inches.
  • Supports a maximum working load of 400 lb.
  • Meets applicable OSHA, ANSI, and CSA standards.
  • Suitable for linemen, miners, utility, or construction work.
Catalog No. 50-005-07964 Supplier MSC Supplier No. 66552241
This body belt is designed for work positioning, featuring a single D-ring, X-Large size, and 1-3/4 inch nylon webbing with a tongue buckle. It is suitable for waist sizes from 42 to 52 inches and supports a maximum working load of 400 lb, meeting OSHA, ANSI, and CSA standards.
  • Designed for work positioning only.
  • Features a single D-ring.
  • Constructed with durable nylon webbing.
  • Equipped with a tongue buckle.
  • X-Large size, fitting waist sizes from 42 to 52 inches.
  • Webbing width of 1-3/4 inches.
  • Supports a maximum working load of 400 lb.
  • Meets applicable OSHA, ANSI, and CSA standards.
  • Suitable for linemen, miners, utility, or construction work.