Learn More
MSC Honeywell Miller Fall Protection Harness, Universal, 420 lb, Polyester, Back D-Ring, Quick-Connect Buckles
Catalog No. 5000508185
Encompass
This fall protection harness is designed for general use, offering enhanced comfort and safety for workers. It features anatomically and ergonomically designed breathable paddings for all-day wear and greater freedom of movement.
  • Highly breathable design
  • Lightweight construction
  • Ergonomic shoulder and back padding
  • Leg and shoulder webbing
  • Dedicated SRL web link
  • Resettable lanyard keepers
  • Smart anti-tangle solution for hassle-free donning
Catalog No. 50-005-08185 Supplier MSC Supplier No. 28480853
This fall protection harness is designed for general use, offering enhanced comfort and safety for workers. It features anatomically and ergonomically designed breathable paddings for all-day wear and greater freedom of movement.
  • Highly breathable design
  • Lightweight construction
  • Ergonomic shoulder and back padding
  • Leg and shoulder webbing
  • Dedicated SRL web link
  • Resettable lanyard keepers
  • Smart anti-tangle solution for hassle-free donning