Learn More
MSC Honeywell Miller Full Body Harness, AirCore, Tongue Buckle, 3X-Large, Green
Catalog No. 5000507190
Encompass
This premium full-body safety harness offers secure fall protection. It features easy-to-use tongue buckle adjustments and breathable AirCore webbing for a customized, comfortable fit. Multiple D-rings provide versatile anchorage, and its lightweight design ensures all-day comfort. Suitable for various elevated work applications, this harness meets ANSI Z359 fall protection standards and supports OSHA compliance.
  • Secure fall protection with premium full-body design.
  • Easy-to-use tongue buckle adjustments for a customized fit.
  • Breathable AirCore webbing and lightweight construction for comfort.
  • Multiple D-rings provide versatile anchorage points.
  • 5 points of adjustment for a secure fit.
  • High-strength, breathable webbing with elastic components.
  • 3X-Large sizing for enhanced comfort and mobility during long tasks.
Catalog No. 50-005-07190 Supplier MSC Supplier No. 80890627
This premium full-body safety harness offers secure fall protection. It features easy-to-use tongue buckle adjustments and breathable AirCore webbing for a customized, comfortable fit. Multiple D-rings provide versatile anchorage, and its lightweight design ensures all-day comfort. Suitable for various elevated work applications, this harness meets ANSI Z359 fall protection standards and supports OSHA compliance.
  • Secure fall protection with premium full-body design.
  • Easy-to-use tongue buckle adjustments for a customized fit.
  • Breathable AirCore webbing and lightweight construction for comfort.
  • Multiple D-rings provide versatile anchorage points.
  • 5 points of adjustment for a secure fit.
  • High-strength, breathable webbing with elastic components.
  • 3X-Large sizing for enhanced comfort and mobility during long tasks.