Learn More
MSC Honeywell Miller Full Body Harness, Back D-Ring, S/M, Mating Chest/Tongue Leg Buckles, 140 kg Capacity
Catalog No. 5000508286
Encompass
The H500 Industry Standard Full-Body Harness offers essential fall protection with maximum comfort and freedom of movement. Its innovative design prevents webbing tangling and bunching, allowing users to concentrate on tasks without restrictive equipment. This safety harness synthesizes safety, quality, and comfort, keeping workers safe and supported.
  • Quick-connect chest and leg buckles for a secure, reliable fit.
  • Smart anti-tangle design and water repellent webbing.
  • Breathable shoulder/back padding, anatomically designed for pressure relief.
  • Side-adjustment straps for better ergonomic fit.
  • Horizontal leg design enhances user comfort.
  • Dedicated SRL web link.
  • User-friendly, resettable pull-free lanyard keepers.
  • Two clearly identifiable fall impact indicators.
  • Rated 140 kg max weight capacity.
Catalog No. 50-005-08286 Supplier MSC Supplier No. 12417937
The H500 Industry Standard Full-Body Harness offers essential fall protection with maximum comfort and freedom of movement. Its innovative design prevents webbing tangling and bunching, allowing users to concentrate on tasks without restrictive equipment. This safety harness synthesizes safety, quality, and comfort, keeping workers safe and supported.
  • Quick-connect chest and leg buckles for a secure, reliable fit.
  • Smart anti-tangle design and water repellent webbing.
  • Breathable shoulder/back padding, anatomically designed for pressure relief.
  • Side-adjustment straps for better ergonomic fit.
  • Horizontal leg design enhances user comfort.
  • Dedicated SRL web link.
  • User-friendly, resettable pull-free lanyard keepers.
  • Two clearly identifiable fall impact indicators.
  • Rated 140 kg max weight capacity.