Learn More
Enterprise Technology Solutions Honeywell N95 Respirator Masks,Cone,Disposable,20/BX,WE
SDP
Catalog No. 5000828319
Encompass_Preferred
Designed to fit most face shapes and sizes, this Molded Cup N95 Respirator Mask maximizes your protection against hazardous airborne irritants. Proprietary filter media features electrostatic, nonwoven, polypropylene polyester fibers to reduce breathing resistance and ensure proper filtration. Form-fitted design is contoured to minimize pressure points and fit a wide variety of face shapes. Molded nose bridge is compatible with most eyewear. Dual, welded head straps keep the safety mask firmly in place. Straps also feature a woven construction to help prevent breakage and hair entanglement. Durable, molded outer shell prevents the breathing mask from collapsing in moist and humid environments. Lightweight and latex-free, this disposable mask offers a 95 percent filtration efficiency against solid and liquid particles (excluding oils), making it ideal for sanding and working with materials like fiberglass and insulation. Mask is recommended by the CDC for pandemic protection.
Catalog No. 50-008-28319 Supplier Enterprise Technology Solutions Supplier No. HWLDC300N95
Designed to fit most face shapes and sizes, this Molded Cup N95 Respirator Mask maximizes your protection against hazardous airborne irritants. Proprietary filter media features electrostatic, nonwoven, polypropylene polyester fibers to reduce breathing resistance and ensure proper filtration. Form-fitted design is contoured to minimize pressure points and fit a wide variety of face shapes. Molded nose bridge is compatible with most eyewear. Dual, welded head straps keep the safety mask firmly in place. Straps also feature a woven construction to help prevent breakage and hair entanglement. Durable, molded outer shell prevents the breathing mask from collapsing in moist and humid environments. Lightweight and latex-free, this disposable mask offers a 95 percent filtration efficiency against solid and liquid particles (excluding oils), making it ideal for sanding and working with materials like fiberglass and insulation. Mask is recommended by the CDC for pandemic protection.