Learn More
MSC Honeywell Nitrile Chemical Gloves, Small, Double Coated, 1.7 mm, A3/C Cut Resistance
Catalog No. 5000509541
Encompass
These nitrile chemical gloves are designed for demanding chemical environments, providing high chemical protection, reliable cut resistance, superior wet and dry grip, enhanced dexterity, and improved comfort for the user.
  • Provides advanced chemical protection.
  • Offers reliable A3/C cut resistance.
  • Features enhanced wet and dry grip for secure handling.
  • Integrated cut liner provides better tactility and dexterity.
  • Moisture dispersion technology keeps hands dry.
  • Ergonomic design reduces hand fatigue.
  • Compliant with food contact legislation.
  • Silicone-free for diverse applications.
Catalog No. 50-005-09541 Supplier MSC Supplier No. 18362996
These nitrile chemical gloves are designed for demanding chemical environments, providing high chemical protection, reliable cut resistance, superior wet and dry grip, enhanced dexterity, and improved comfort for the user.
  • Provides advanced chemical protection.
  • Offers reliable A3/C cut resistance.
  • Features enhanced wet and dry grip for secure handling.
  • Integrated cut liner provides better tactility and dexterity.
  • Moisture dispersion technology keeps hands dry.
  • Ergonomic design reduces hand fatigue.
  • Compliant with food contact legislation.
  • Silicone-free for diverse applications.