Learn More
MSC Honeywell North Coupler Assembly, Hansen Style, 3/8 in ID Hose, CF2000 Series
Catalog No. 5000509405
Encompass
This coupler assembly features one male and one female quick-disconnect coupler, designed for secure and efficient connection of breathing tubes to 3/8 inch ID hoses. It is specifically compatible with CF2000 Series for use with supplied air respirators.
  • Assembly with one male and one female coupler
  • Connects breathing tube to 3/8 inch ID hose
  • Compatible with CF2000 series
  • 2 inch length
  • Hansen style quick-disconnect
Catalog No. 50-005-09405 Supplier MSC Supplier No. 48464861
This coupler assembly features one male and one female quick-disconnect coupler, designed for secure and efficient connection of breathing tubes to 3/8 inch ID hoses. It is specifically compatible with CF2000 Series for use with supplied air respirators.
  • Assembly with one male and one female coupler
  • Connects breathing tube to 3/8 inch ID hose
  • Compatible with CF2000 series
  • 2 inch length
  • Hansen style quick-disconnect