Learn More
MSC Honeywell North Emergency Escape Mouthbit Respirator, Ammonia Protection, Silicone Mouthpiece, Universal Size
Catalog No. 5000507523
Encompass
This emergency escape mouthbit respirator is designed for use where ammonia may be present, such as in refrigeration facilities. It provides a quick and economical method to escape non-IDLH emergency situations, suitable for visitors or as a safety precaution for employees against potential chemical leaks.
  • Comfortable silicone mouthpiece, hypoallergenic and latex-free.
  • Wider opening mouthpiece for a natural feel.
  • Soft textured mouthpiece prevents slipping.
  • Removable mouthpiece for periodic cleaning.
  • Large, soft nose pads ensure secure, comfortable fit.
  • Wider nose clip fits various facial types.
  • Nylon neck strap prevents loss during escape.
  • Nylon carrying bag protects the respirator.
  • Belt loops for constant accessibility.
  • Zip lock storage bag for emergency access and daily inspection.
Catalog No. 50-005-07523 Supplier MSC Supplier No. 85776474
This emergency escape mouthbit respirator is designed for use where ammonia may be present, such as in refrigeration facilities. It provides a quick and economical method to escape non-IDLH emergency situations, suitable for visitors or as a safety precaution for employees against potential chemical leaks.
  • Comfortable silicone mouthpiece, hypoallergenic and latex-free.
  • Wider opening mouthpiece for a natural feel.
  • Soft textured mouthpiece prevents slipping.
  • Removable mouthpiece for periodic cleaning.
  • Large, soft nose pads ensure secure, comfortable fit.
  • Wider nose clip fits various facial types.
  • Nylon neck strap prevents loss during escape.
  • Nylon carrying bag protects the respirator.
  • Belt loops for constant accessibility.
  • Zip lock storage bag for emergency access and daily inspection.