Learn More
MSC Honeywell North Finger Cots, Natural Rubber, Powder-Free, X-Large, 3 mil
Catalog No. 5000507314
Encompass
These natural rubber finger cots are designed for use in industrial and food-grade environments. They are powder-free, white, and feature tapered fingertips to promote high tactile sensitivity, making them suitable for various applications.
  • Powder-free
  • Industrial food grade
  • White color
  • X-Large size
  • 2-3/4 in long
  • 3 mil thick
  • Rolled cuff style
  • Dipped construction
  • Tapered fingertips for high tactile sensitivity
Catalog No. 50-005-07314 Supplier MSC Supplier No. 78656113
These natural rubber finger cots are designed for use in industrial and food-grade environments. They are powder-free, white, and feature tapered fingertips to promote high tactile sensitivity, making them suitable for various applications.
  • Powder-free
  • Industrial food grade
  • White color
  • X-Large size
  • 2-3/4 in long
  • 3 mil thick
  • Rolled cuff style
  • Dipped construction
  • Tapered fingertips for high tactile sensitivity