Learn More
MSC Honeywell North Respirator Cartridge, Ammonia/Methylamine Protection, 7700 Series, Green
Catalog No. 5000508562
Encompass
This respirator cartridge provides protection against ammonia and methylamine vapors. It features a green color code and is compatible with 7700 series half masks and full facepieces, ensuring reliable respiratory protection in hazardous environments.
  • Protection against ammonia vapors
  • Protection against methylamine vapors
  • Green color code
  • Compatible with 7700, 5500, RU8500, RU8800 series half masks
  • Compatible with 5400, RU6500, 7600 series full facepieces
  • Low profile design helps prevent catching on objects
  • Easy user seal checks
Catalog No. 50-005-08562 Supplier MSC Supplier No. 06558324
This respirator cartridge provides protection against ammonia and methylamine vapors. It features a green color code and is compatible with 7700 series half masks and full facepieces, ensuring reliable respiratory protection in hazardous environments.
  • Protection against ammonia vapors
  • Protection against methylamine vapors
  • Green color code
  • Compatible with 7700, 5500, RU8500, RU8800 series half masks
  • Compatible with 5400, RU6500, 7600 series full facepieces
  • Low profile design helps prevent catching on objects
  • Easy user seal checks