Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Perfect-Coat™ Latex-Coated Medium Weight Thermal-Lined Gloves

Catalog No. 19014388b
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-014-388B Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Warning: These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-014-388B Supplier Honeywell™ Supplier No. 451PCM
This item is not returnable. View return policy
Warning: These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Heavyweight thermal liner on the inside of the glove provides warmth, while the natural rubber coating enhances grip capabilities.

  • Heavyweight thermal knit liner is napped on the inside for warmth
  • Cotton/Polyester blend helps to wick-away moisture
  • Knit liner is soft and comfortable
  • Natural rubber palm and fingertip coating
  • Rubber coating provides excellent grip and flexibility
TRUSTED_SUSTAINABILITY

Specifications

Coated Area Palm, Fingers
Coating Material Latex
Color Gray
Exterior Finish Partially Textured
Hand Specific
Material Polycotton
Size Medium
For Use With (Application) Cold Storage, Construction, Landscaping, General Maintenance, and Material Handling
Glove Cuff Style Knit
Liner Material Thermal
Series 451PC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.